Learn More
Electron Microscopy Sciences Dietrich's Fixative 100 ML
SDP
Catalog No. 501901169
Encompass_Preferred
Prepared fresh upon receipt of order to ensure maximum shelf life. Ingredients are Ethanol, formalin, glacial acetic acid, distilled water.
Catalog No. 50-190-1169 Supplier Electron Microscopy Sciences Supplier No. 1595820
Prepared fresh upon receipt of order to ensure maximum shelf life. Ingredients are Ethanol, formalin, glacial acetic acid, distilled water.