Learn More
Electron Microscopy Sciences Dietrich's Fixative Pre-filled Vials 10 ML
SDP
Catalog No. 501901168
Encompass_Preferred
Prepared fresh upon receipt of order to ensure maximum shelf life. Ingredients are Ethanol, formalin, glacial acetic acid, distilled water. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
Catalog No. 50-190-1168 Supplier Electron Microscopy Sciences Supplier No. 1595810
Prepared fresh upon receipt of order to ensure maximum shelf life. Ingredients are Ethanol, formalin, glacial acetic acid, distilled water. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.