Learn More
Newcomer Supply Thiazine Stain, 1 gallon
SDP
Catalog No. NC9801931
Encompass
Solution B of the Differential Stain Kit. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9801931 Supplier Newcomer Supply Supplier No. 10522B
Solution B of the Differential Stain Kit. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.