Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Noble Agar

Catalog No. DF0142152 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0142-15-2 100 g
DF0142-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DF0142-15-2 Supplier BD Supplier No. 214220
This item is not returnable. View return policy

Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.

Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.

 

  • Solidifying agent
  • Free of impurities

 

Applications:

  • Used for electrophoretic and nutritional procedures
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Content And Storage <30°C
Additive Noble Agar
Form Dehydrated
For Use With (Application) Preparing microbiological culture media, for Electrophoretic and nutritional procedures
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.