Learn More
Medchemexpress LLC Difelikefalin-d5 hydrochloride | 97.6% | 794.26 | 5 MG
SDP
Catalog No. 5000387348
Encompass_Preferred
Difelikefalin-d5 hydrochloride is the deuterium-labeled Difelikefalin (HY-17609). Difelikefalin is a peripherally restricted and selective agonist of the kappa opioid receptor (KOR). It produces anti-inflammatory effects and may modulate pruritus in conditions such as chronic kidney disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-87348 Supplier Medchemexpress LLC Supplier No. HY17609S15MG
Difelikefalin-d5 hydrochloride is the deuterium-labeled Difelikefalin (HY-17609). Difelikefalin is a peripherally restricted and selective agonist of the kappa opioid receptor (KOR). It produces anti-inflammatory effects and may modulate pruritus in conditions such as chronic kidney disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs