Learn More
Medchemexpress LLC Difelikefalin-d5 hydrochloride (CR-845-d5 hydrochloride) | 1024828-77-0 | 97.6% | 794.26 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17609S11MG
Encompass_Preferred
Difelikefalin-d5 (CR-845-d5) hydrochloride is the deuterium-labeled Difelikefalin. It acts as a peripherally restricted and selective agonist of the kappa opioid receptor (KOR), demonstrating anti-inflammatory effects and potentially modulating pruritus in conditions such as chronic kidney disease.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87218