Learn More
Medchemexpress LLC Difenoconazole (Standard) | 119446-68-3 | 95.55% | 100 MG
SDP
Catalog No. 5000409406
Encompass_Preferred
Difenoconazole (Standard) is the analytical standard of Difenoconazole, a sterol demethylation inhibitor acting as a fungicide. It interferes with mycelial growth and inhibits spore germination by binding to the heme portion of fungal cytochrome P450 51. This product is intended for research and analytical applications.
  • Analytical standard grade
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-09406 Supplier Medchemexpress LLC Supplier No. HYB0850R100MG
Difenoconazole (Standard) is the analytical standard of Difenoconazole, a sterol demethylation inhibitor acting as a fungicide. It interferes with mycelial growth and inhibits spore germination by binding to the heme portion of fungal cytochrome P450 51. This product is intended for research and analytical applications.
  • Analytical standard grade
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications