Learn More
Medchemexpress LLC Difenoconazole (Standard) | 119446-68-3 | 95.55% | 25 MG
SDP
Catalog No. 5000450500
Encompass_Preferred
Difenoconazole (Standard) is the analytical standard of Difenoconazole, intended for research and analytical applications. This product acts as a sterol demethylation inhibitor and a fungicide, binding to the heme portion of fungal cytochrome P450 51 to interfere with mycelial growth and inhibit spore germination. It is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Sterol demethylation inhibitor
  • Functions as a fungicide
  • Interferes with mycelial growth
  • Inhibits spore germination of pathogens
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-50500 Supplier Medchemexpress LLC Supplier No. 25MGHYB0850R
Difenoconazole (Standard) is the analytical standard of Difenoconazole, intended for research and analytical applications. This product acts as a sterol demethylation inhibitor and a fungicide, binding to the heme portion of fungal cytochrome P450 51 to interfere with mycelial growth and inhibit spore germination. It is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Sterol demethylation inhibitor
  • Functions as a fungicide
  • Interferes with mycelial growth
  • Inhibits spore germination of pathogens
  • Used in HPLC, GC, and MS experiments