Learn More
Zen Bio Inc DIFF MEDIA/PRE ADIPOCYTE 100ML
SDP
Catalog No. NC9860641
Encompass
Differentiation Medium , 100ml/bottle , used to stimulate the differentiation of cultured preadipocytes into adipocytes, Contents: Adipocyte Medium;0.25 mM isobutyl methylxanthine;PPAR agonist
Catalog No. NC9860641 Supplier Zen Bio Inc Supplier No. DM2
Differentiation Medium , 100ml/bottle , used to stimulate the differentiation of cultured preadipocytes into adipocytes, Contents: Adipocyte Medium;0.25 mM isobutyl methylxanthine;PPAR agonist