Learn More
Siemens Siemens Healthcare Shop Dashboard
Catalog No. NC1437686
Encompass_Preferred
The Siemens Healthineers Clinitek Status+ Analyzer is an in vitro diagnostic urine chemistry analyzer that provides quantitative test results for urine chemistry parameters including glucose, bilirubin, ketones, specific gravity, blood, pH, protein, urobilinogen, nitrite, and leukocytes. It utilizes dry-slide technology with test strips, has a throughput of up to 100 samples per hour, and offers features like continuous loading, auto-calibration, and result storage. The compact tabletop analyzer has minimal maintenance requirements.
This item is not returnable. View return policy
Catalog No. NC1437686 Supplier Siemens Supplier No. 10736132
The Siemens Healthineers Clinitek Status+ Analyzer is an in vitro diagnostic urine chemistry analyzer that provides quantitative test results for urine chemistry parameters including glucose, bilirubin, ketones, specific gravity, blood, pH, protein, urobilinogen, nitrite, and leukocytes. It utilizes dry-slide technology with test strips, has a throughput of up to 100 samples per hour, and offers features like continuous loading, auto-calibration, and result storage. The compact tabletop analyzer has minimal maintenance requirements.