Learn More
Electron Microscopy Sciences Differential Quick Stain Kit 500 ML
SDP
Catalog No. NC0851891
Encompass_Preferred
Modified Giemsa Stain kit is for quick turn around results. The kit may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms. The kit comes compete with a fixative for air dried cell suspensions and or touch preparation slides. Applications include fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies. This stain offers results in 15 seconds. The Kit comes complete with 3 solutions.
Catalog No. NC0851891 Supplier Electron Microscopy Sciences Supplier No. 2609650
Modified Giemsa Stain kit is for quick turn around results. The kit may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms. The kit comes compete with a fixative for air dried cell suspensions and or touch preparation slides. Applications include fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies. This stain offers results in 15 seconds. The Kit comes complete with 3 solutions.