Learn More
Newcomer Supply Xanthene Stain, 1 gallon
SDP
Catalog No. NC9001099
Encompass
Solution A of the Differential Stain Kit. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9001099 Supplier Newcomer Supply Supplier No. 10521B
Solution A of the Differential Stain Kit. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.