Learn More
IMEB, Inc DIFF STAIN KIT Includes 8 oz. Bottle Solution A (Fixative), 8 oz. Bottle Solution B (Azure Stain), 8 oz. Bottle Solution C (Xanthene dyes) and 3- 2 oz. Staining Containers and 1 Storage Tray
SDP
Catalog No. NC9979740
Encompass_Preferred
Diff Stain kit is used to provide Nuclear and cytoplasmic staining and recently has provided excellent staining for Helicobacter Pylori. The result is Nuclear staining in blue to purple staining and pink to orange with paraffin and frozen sections. Helicobacter pylori provides a deep rich purple. This stain is also excellent as a Hematology stain. Contains: 8 oz. Bottle solution A (Fixative), 8 oz. Bottle solution B (Azure Stain), 8 oz. Bottle solution C (Xanthene dyes) and 3- 2 oz. Staining containers and 1- storage tray.
Catalog No. NC9979740 Supplier IMEB, Inc Supplier No. K7128
Diff Stain kit is used to provide Nuclear and cytoplasmic staining and recently has provided excellent staining for Helicobacter Pylori. The result is Nuclear staining in blue to purple staining and pink to orange with paraffin and frozen sections. Helicobacter pylori provides a deep rich purple. This stain is also excellent as a Hematology stain. Contains: 8 oz. Bottle solution A (Fixative), 8 oz. Bottle solution B (Azure Stain), 8 oz. Bottle solution C (Xanthene dyes) and 3- 2 oz. Staining containers and 1- storage tray.