Learn More
MODULUS DATA SYSTEMS, INC. DIFFCOUNT™ Manual Differential Cell Counter
Catalog No. 21403167
Change view
Click to view available options
Voltage:
100/240 V
Catalog No. Voltage
21-403-167 100/240 V
1 options
Catalog No. 21-403-167 Supplier MODULUS DATA SYSTEMS, INC. Supplier No. 10412M
Add to Cart
Add to Cart
Next generation digital manual differential counter with touchscreen and customizable lab features.
  • Use familiar 8 or 12-channel quick-start modes for easy transition to our newest generation DIFFCOUNT™, or set up custom users, modes, and comments based on your lab's unique needs.
  • Modern touchscreen interface provides intuitive navigation and control for efficient operation.
  • On-screen, dynamic, user-defined channel names that change with each mode (no physical overlay swaps).
  • Absolute calculations including WBC with nRBC correction for accurate results.
  • Count totals for all channels, individual channels and multiple channel subgroups concurrently with individual percentages.
  • Set up custom comment fields (descriptive and semi-quantitative) for your facility's preferred nomenclature such as cell type morphology and frequency.
  • Quick select one-touch access to common user workflows with simplified interaction through the touchscreen display.
  • Set user, mode or lab-specific audio feedback and lockout controls to ensure consistent operation.
  • Barcode or QR input supported with compatible scanner (sold separately, P/N 14500) for streamlined data entry.
  • Print to our compatible 80 mm thermal ticket printer (sold separately, P/N 24700) or to PostScript-compatible desktop printers via USB for flexible output options.
  • Import and export user-customized configuration and settings for backup and easy replication to additional DIFFCOUNT™ counters (10 to 412 M models only).
  • Extensive user customization options maintain familiarity for users of earlier models while enhancing usability.
Specifications
Product Type Manual Counter
Display Type TFT Color LCD
Voltage 100/240 V