Learn More
Modulus Data Systems Inc. DIFFCOUNT™ Thermal Printer
Catalog No. 21402597
Change view
Click to view available options
For Use With (Equipment):
DIFFCOUNT 10-412 12-Channel Differential Cell Counter
Catalog No. For Use With (Equipment)
21-402-597 DIFFCOUNT 10-412 12-Channel Differential Cell Counter
1 options
Catalog No. 21-402-597 Supplier Modulus Data Systems Inc. Supplier No. 24700
Add to Cart
Add to Cart
Complete your benchtop cell counting workstation with the DIFFCOUNT Ticket Printer, for use with the DIFFCOUNT 10-412 12-Channel Differential Cell Counter.
  • Print counts, calculations, and comments to this compact benchtop ticket printer for clear, easy-to-read and accurate results.
  • Each printer comes with a single starter roll of BPA-free thermal ticket paper and uses standard 80 mm refills (Sold separately).
  • One-year warranty.
Specifications
Type Printer
For Use With (Equipment) DIFFCOUNT 10-412 12-Channel Differential Cell Counter
Voltage 110/220 V