Learn More
Avanti Polar Lipids DIFFERENTIAL ION MOBILITY SYS
Catalog No. NC1937471
Encompass
Differential Ion Mobility System Suitability Synthetic Standard Mixture. Mixture contains 1mg/ml of each lipid below (0.25mg/ml PI), 0.5ml per vial in chloroform: methanol (1:1). We recommend purchasing Bovine Heart Extract as an accompanying product.
Catalog No. NC1937471 Supplier Avanti Polar Lipids Supplier No. 791500X1EA
Differential Ion Mobility System Suitability Synthetic Standard Mixture. Mixture contains 1mg/ml of each lipid below (0.25mg/ml PI), 0.5ml per vial in chloroform: methanol (1:1). We recommend purchasing Bovine Heart Extract as an accompanying product.