Learn More
Newcomer Supply Differential Stain Kit, 500 ml kit
SDP
Catalog No. NC9535624
Encompass
Newcomer Supply Differential Stain Kit, a modification of the Wright Giemsa Stain, uses a methanol fixative and aqueous based stains to provide a rapid 3-step process for differential assessment of:  peripheral blood smears, touch imprints, fine needle aspirations (FNA), bone marrow biopsy aspirations, and detecting microorganisms. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9535624 Supplier Newcomer Supply Supplier No. 9112B
Newcomer Supply Differential Stain Kit, a modification of the Wright Giemsa Stain, uses a methanol fixative and aqueous based stains to provide a rapid 3-step process for differential assessment of:  peripheral blood smears, touch imprints, fine needle aspirations (FNA), bone marrow biopsy aspirations, and detecting microorganisms. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.