Learn More
Electron Microscopy Sciences Differentiating Solution 500 ML
SDP
Catalog No. 5031923
Encompass_Preferred
Stain DNA & RNA. References: Taft, E.B., Stain Tech., 26:205-212, 1951.
Catalog No. 50-319-23 Supplier Electron Microscopy Sciences Supplier No. 2677702
Stain DNA & RNA. References: Taft, E.B., Stain Tech., 26:205-212, 1951.