Learn More
Lonza Walkersville hMSC Human Mesenchymal Stem Cell Chondrogenic Differentiation Basal Medium
Catalog No. NC9925487
Encompass
Stem cells possess the unique ability to differentiate into multiple lineages of cells including chondrocytes. Lonza’s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation, with the addition of TGF-β3, of human bone marrow derived mesenchymal stem cells into mature, functionally active chondrocytes in as few as 14 days. Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin. Partial List of Cells Differentiated with hMSC Chondrogenic Differentiation BulletKit Medium*: Human bone marrow derived stem cells Human umbilical cord blood derived stem cells Human umbilical cord matrix derived stem cells C57BL/6 mice bone marrow derived stem cells human adipose derived stem cells Human hamstring tendon derived stem cells.
Catalog No. NC9925487 Supplier Lonza Walkersville Supplier No. PT3925
Stem cells possess the unique ability to differentiate into multiple lineages of cells including chondrocytes. Lonza’s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation, with the addition of TGF-β3, of human bone marrow derived mesenchymal stem cells into mature, functionally active chondrocytes in as few as 14 days. Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin. Partial List of Cells Differentiated with hMSC Chondrogenic Differentiation BulletKit Medium*: Human bone marrow derived stem cells Human umbilical cord blood derived stem cells Human umbilical cord matrix derived stem cells C57BL/6 mice bone marrow derived stem cells human adipose derived stem cells Human hamstring tendon derived stem cells.