Learn More
AdipoGen Differolide
SDP
Supplier:  AdipoGen BVT0401M001
Encompass_Preferred
Chemical. CAS 106750-00-9. Formula C12H12O4. MW 220.2. Isolated from Streptomyces aurantiogriseus. Signal substance. Microbial bioregulator.
Catalog No. 50-256-1945