Learn More
Sigma Aldrich Fine Chemicals Biosciences Diffinity RapidTipR for PC
Catalog No. 502904006
Encompass
Diffinity RapidTip effectively removes dNTPs primers and primer dimers while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The dispensed SOLUTIONon yields purified high quality DNA ready for downstream applications such as DNA sequencing SNP analysis and microarray printing.
Catalog No. 50-290-4006 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D194796RXN
Diffinity RapidTip effectively removes dNTPs primers and primer dimers while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The dispensed SOLUTIONon yields purified high quality DNA ready for downstream applications such as DNA sequencing SNP analysis and microarray printing.