Learn More
Lifeline Cell Technology DIFFKT MSC ADAP LIFELINE KT
SDP
Catalog No. 50311921
Encompass_Preferred
ChondroLife Complete Chondrogenesis Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into chondrocytes. ChondroLife contains no antimicrobials and no phenol red.
Catalog No. 50-311-921 Supplier Lifeline Cell Technology Supplier No. LM0022
ChondroLife Complete Chondrogenesis Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into chondrocytes. ChondroLife contains no antimicrobials and no phenol red.