Learn More
Lifeline Cell Technology DIFFKT MSC ADAP LIFELINE KT
SDP
Catalog No. 50311912
Encompass_Preferred
AdipoLife DfKt-1 Adipogenesis Medium Complete Kit (AdipoLife Basal Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into adipocytes. AdipoLife contains no antimicrobials and no phenol red.
Catalog No. 50-311-912 Supplier Lifeline Cell Technology Supplier No. LL0050
AdipoLife DfKt-1 Adipogenesis Medium Complete Kit (AdipoLife Basal Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into adipocytes. AdipoLife contains no antimicrobials and no phenol red.