Learn More
Lifeline Cell Technology DIFFKT MSC OSTEO LIFELINE KT
SDP
Catalog No. 50311922
Encompass_Preferred
OsteoLife Complete Osteogenesis Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into osteocytes. Contains no antimicrobials and no phenol red that can cause cell stress and masking effects.
Catalog No. 50-311-922 Supplier Lifeline Cell Technology Supplier No. LM0023
Add to Cart
Add to Cart
OsteoLife Complete Osteogenesis Medium 100 mL is optimized for the differentiation of adult human mesenchymal stem cells into osteocytes. Contains no antimicrobials and no phenol red that can cause cell stress and masking effects.