Learn More
Cayman Chemical DIffractaIc AcId 2.5mg
SDP
Catalog No. 502933204
Encompass_Preferred
A lichen metabolite with diverse biological activities; cytotoxic to HCT116, HeLa, and MCF-7 cancer cells (IC50s = 42.2, 64.6, and 93.4 μM, respectively); inhibits growth of M. tuberculosis (MIC = 41.7 μM); reduces indomethacin-induced neutrophil infiltration, lipid peroxidation, MPx activity, and the number of gastric lesions as well as reverses decreases in SOD and GPx activities in rat gastric mucosa from 25-200 mg/kg; reduces acetic acid-induced writhing and increases the pressure pain threshold in mice
Catalog No. 50-293-3204 Supplier Cayman Chemical Supplier No. 242082.5 MG
A lichen metabolite with diverse biological activities; cytotoxic to HCT116, HeLa, and MCF-7 cancer cells (IC50s = 42.2, 64.6, and 93.4 μM, respectively); inhibits growth of M. tuberculosis (MIC = 41.7 μM); reduces indomethacin-induced neutrophil infiltration, lipid peroxidation, MPx activity, and the number of gastric lesions as well as reverses decreases in SOD and GPx activities in rat gastric mucosa from 25-200 mg/kg; reduces acetic acid-induced writhing and increases the pressure pain threshold in mice