Learn More
Medchemexpress LLC Diffractaic acid | 436-32-8 | MFCD00078235 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN23995MG
Encompass_Preferred
Diffractaic acid, a major constituent of *U. longissimi*, acts as an effective proapoptotic agent in various disorders research. It is also the analgesic and antipyretic component of *Usnea diffracta*.
  • Antiproliferative activity against human HaCaT cells (IC50 = 2.6 μM)
  • Inhibition of LTB4 biosynthesis in A-23187-stimulated bovine PMNL cells (IC50 = 8 μM)
  • Shows an analgesic effect in mice by acetic acid-induced writhing and tail-pressure methods
Catalog No. 50-004-20908