Learn More
Vernier™ Diffraction Apparatus
SDPSCI_ED
Catalog No. S16001ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Mapping light intensity
Catalog No. For Use With (Application)
S16001ND Mapping light intensity
1 options
Catalog No. S16001ND Supplier Vernier™ Supplier No. DAK
Add to Cart
Add to Cart
Includes
Diffraction Slit System, Red Diffraction Laser, Combination Linear Position and High Sensitivity Light Sensor, Power Supply
Requires
Requires an interface and software, sold separately
Use the Diffraction Apparatus to map light intensity vs. position for various slit geometries.
Red Laser Wavelength: 635 nm +/- 5 nm (Class 2 laser product) Light Sensor Full Scale Ranges: 1, 10, and 100 μW Linear Position Sensor range: 150 mm Linear Position Sensor resolution: 40 μm Available slits Single Slits (4) 0.02 mm 0.04 mm 0.08 mm 0.16 mm Variable Slits Wedge: 0.02 - 0.2 mm wide Double Slit: 0.04 mm wide, spacing 0.25 - 0.75 mm Double Slits (4 pairs) 0.04 mm wide at 0.125 mm apart 0.04 mm wide at 0.5 mm apart 0.08 mm wide at 0.25 mm apart 0.08 mm wide at 0.5 mm apart Variable Double Slit:Same as variable slits group above Multiple Slits 0.04 mm wide at 0.25 mm apart 4 sets, 2, 3, 4, 5 slits Comparisons: 4 pairs of single/double slits: 0.04 mm single + 0.04/0.25 mm double, doubles 0.04/0.25 +0.04/0.50, doubles 0.04/0.25+0.08/0.25, double 0.04/0.25 + triple, 0.04/0.25
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Mapping light intensity
For Use With (Equipment) Combination Track/Optics Bench