Learn More
Ted Pella Inc Diffraction Grating Replica and Latex Calibration Standard
SDP
Catalog No. NC1264490
Encompass
Two-in-one; this standard for calibration provides a double-check of the accuracy of magnification calibration. It is particularly useful at higher magnification. The latex sphere size is 0.261 µm and the carbon grating replica with Au/Pd shadowing is 2160 lines/mm. Supplied on a G400, 400 square mesh copper Gilder grid.
Catalog No. NC1264490 Supplier Ted Pella Inc Supplier No. 603
Two-in-one; this standard for calibration provides a double-check of the accuracy of magnification calibration. It is particularly useful at higher magnification. The latex sphere size is 0.261 µm and the carbon grating replica with Au/Pd shadowing is 2160 lines/mm. Supplied on a G400, 400 square mesh copper Gilder grid.