Learn More
Perkin Elmer US LLC Diffuse Reflectance Accessory for the Spectrum Two
Catalog No. 502117213
Encompass_Preferred
The accessory requires no alignment and allows for macro or micro sampling. It is compatible with Spectrum Two spectrometers.
Catalog No. 50-211-7213 Supplier Perkin Elmer US LLC Supplier No. L1600111
Add to Cart
Add to Cart
Request a Quote
The accessory requires no alignment and allows for macro or micro sampling. It is compatible with Spectrum Two spectrometers.