Learn More
Grainger Atlantic Diffusers Disc Diffuser, Coarse Bubble, 20 Scfm, 32 Sq In.
Catalog No. 502792080
Encompass_Preferred
This disc-style diffuser is designed for efficient aeration in wastewater treatment and oxygen transfer applications. Featuring a coarse bubble design, it delivers consistent performance while utilizing an EPDM or silicone membrane that prevents water backflow and resists clogging.
  • Coarse bubble type with 9 to 12 mm bubble size
  • 32 sq in. active surface area
  • Maximum air flow of 20 scfm
  • Operating range of 3.0 to 20.0 scfm
  • 5 in. diameter with 3/4 in. MPT connection
Catalog No. 50-279-2080 Supplier Grainger Supplier No. 16G397
This disc-style diffuser is designed for efficient aeration in wastewater treatment and oxygen transfer applications. Featuring a coarse bubble design, it delivers consistent performance while utilizing an EPDM or silicone membrane that prevents water backflow and resists clogging.
  • Coarse bubble type with 9 to 12 mm bubble size
  • 32 sq in. active surface area
  • Maximum air flow of 20 scfm
  • Operating range of 3.0 to 20.0 scfm
  • 5 in. diameter with 3/4 in. MPT connection