Learn More
Grainger Atlantic Diffusers Disc Diffuser, Medium Bubble, Polypropylene, 5 in. Diameter, 1 ea.
Catalog No. 502792076
Encompass_Preferred
This medium bubble disc diffuser is designed for optimal performance in water aeration, oxygenation, and wastewater treatment systems. Constructed with a durable polypropylene frame and an EPDM membrane, the unit provides consistent air distribution while minimizing the risk of clogging or fouling. The membrane automatically closes over the air inlet when the air supply is shut down to prevent backflow into the air line.
  • 5 in. disc diameter with 32 sq. in. active surface area
  • Medium bubble size range of 6 mm to 10 mm
  • Airflow requirements of 2.0 to 12.0 scfm
  • Equipped with a 3/4 in. MPT fitting
  • Quick and efficient membrane replacements
  • Compatible with filtered water, oxygen, and wastewater applications
Catalog No. 50-279-2076 Supplier Grainger Supplier No. 16G398
This medium bubble disc diffuser is designed for optimal performance in water aeration, oxygenation, and wastewater treatment systems. Constructed with a durable polypropylene frame and an EPDM membrane, the unit provides consistent air distribution while minimizing the risk of clogging or fouling. The membrane automatically closes over the air inlet when the air supply is shut down to prevent backflow into the air line.
  • 5 in. disc diameter with 32 sq. in. active surface area
  • Medium bubble size range of 6 mm to 10 mm
  • Airflow requirements of 2.0 to 12.0 scfm
  • Equipped with a 3/4 in. MPT fitting
  • Quick and efficient membrane replacements
  • Compatible with filtered water, oxygen, and wastewater applications