Learn More
Cayman Chemical DIflorason DIacetate 10mg
SDP
Catalog No. 502994833
Encompass_Preferred
A corticosteroid; reduces production of the inflammatory cytokines Il-6, Il-4, and Mip-2 and completely inhibits swelling in the EArs of toluene-2,4-diisocyaniate-sensitized mice following topical administration of 20 µL of a 0.05% solution; inhibits RANTES and TARC in a dinitrochlorobenzene-induced mouse model of allergic contact dermatitis; inhibits proliferation of keratinocytes and reduces epidermal thickening in a mouse model of imiquimod-induced psoriasis
Catalog No. 50-299-4833 Supplier Cayman Chemical Supplier No. 2380810 MG
Add to Cart
Add to Cart
A corticosteroid; reduces production of the inflammatory cytokines Il-6, Il-4, and Mip-2 and completely inhibits swelling in the EArs of toluene-2,4-diisocyaniate-sensitized mice following topical administration of 20 µL of a 0.05% solution; inhibits RANTES and TARC in a dinitrochlorobenzene-induced mouse model of allergic contact dermatitis; inhibits proliferation of keratinocytes and reduces epidermal thickening in a mouse model of imiquimod-induced psoriasis