Learn More
Medchemexpress LLC Diflorasone diacetate | 33564-31-7 | MFCD00079159 | 99.9% | 494.52 g·mol⁻1 | C26H32F2O7 | 50 MG
SDP
Catalog No. 5000242792
Encompass_Preferred
Diflorasone diacetate is a synthetic topical corticosteroid supplied as a solid research-grade compound for preclinical, formulation, and analytical applications. It is used to model corticosteroid-responsive dermatoses and to support assay development involving anti-inflammatory steroid activity.
  • Solid powder form suitable for weighing and formulation.
  • High purity (99.86%) for reproducible results.
  • Known molecular weight 494.52 g·mol⁻1 and formula C26H32F2O7.
  • Available in multiple pack sizes for scale flexibility.
  • Suitable for topical and dermatological research studies.
Catalog No. 50-002-42792 Supplier Medchemexpress LLC Supplier No. HY10796150MG
Diflorasone diacetate is a synthetic topical corticosteroid supplied as a solid research-grade compound for preclinical, formulation, and analytical applications. It is used to model corticosteroid-responsive dermatoses and to support assay development involving anti-inflammatory steroid activity.
  • Solid powder form suitable for weighing and formulation.
  • High purity (99.86%) for reproducible results.
  • Known molecular weight 494.52 g·mol⁻1 and formula C26H32F2O7.
  • Available in multiple pack sizes for scale flexibility.
  • Suitable for topical and dermatological research studies.