Learn More
Medchemexpress LLC Diflorasone diacetate | 33564-31-7 | MFCD00079159 | 99.9% | 494.52 g·mol⁻¹ | C26H32F2O7 | 100 MG
SDP
Catalog No. 5000242436
Encompass_Preferred
Diflorasone diacetate is a potent topical corticosteroid supplied for research use in solid form or as a DMSO solution. It is used in pharmacological and dermatological studies to model corticosteroid effects and to evaluate treatments for corticosteroid-responsive skin disorders.
  • Potent anti-inflammatory corticosteroid for topical application studies.
  • Available as solid powder and as a 10 mM solution in DMSO for flexible dosing.
  • High purity suitable for analytical and pharmacological research.
  • Molecular formula C26H32F2O7 and molecular weight 494.52 g·mol⁻¹.
  • Commonly used to evaluate corticosteroid-responsive dermatoses in vitro.
Catalog No. 50-002-42436 Supplier Medchemexpress LLC Supplier No. HY107961100MG
Diflorasone diacetate is a potent topical corticosteroid supplied for research use in solid form or as a DMSO solution. It is used in pharmacological and dermatological studies to model corticosteroid effects and to evaluate treatments for corticosteroid-responsive skin disorders.
  • Potent anti-inflammatory corticosteroid for topical application studies.
  • Available as solid powder and as a 10 mM solution in DMSO for flexible dosing.
  • High purity suitable for analytical and pharmacological research.
  • Molecular formula C26H32F2O7 and molecular weight 494.52 g·mol⁻¹.
  • Commonly used to evaluate corticosteroid-responsive dermatoses in vitro.