Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diflorasone diacetate | 33564-31-7 | MFCD00079159 | 99.9% | 494.52 g/mol | C26H32F2O7 | 25 MG
SDP

Catalog No. 5000242151
Encompass_Preferred

Diflorasone diacetate is a fluorinated corticosteroid research compound (CAS 33564-31-7) used as a topical anti-inflammatory agent in dermatological and inflammation studies. Supplied as a high-purity white to off-white solid, it is intended for laboratory research and formulation applications.

  • High purity (99.9%) solid suitable for analytical and biological studies.
  • Molecular formula C26H32F2O7; molecular weight 494.52 g/mol.
  • Recommended solvent: DMSO for preparing concentrated stock solutions (100 mg/mL).
  • Storage: sealed, away from moisture and light; solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Common applications include topical anti-inflammatory and dermatological research.
  • Available in small quantities suitable for research and reference standard use.

Catalog No. 50-002-42151 Supplier Medchemexpress LLC Supplier No. HY10796125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diflorasone diacetate is a fluorinated corticosteroid research compound (CAS 33564-31-7) used as a topical anti-inflammatory agent in dermatological and inflammation studies. Supplied as a high-purity white to off-white solid, it is intended for laboratory research and formulation applications.

  • High purity (99.9%) solid suitable for analytical and biological studies.
  • Molecular formula C26H32F2O7; molecular weight 494.52 g/mol.
  • Recommended solvent: DMSO for preparing concentrated stock solutions (100 mg/mL).
  • Storage: sealed, away from moisture and light; solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • Common applications include topical anti-inflammatory and dermatological research.
  • Available in small quantities suitable for research and reference standard use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.