Learn More
Medchemexpress LLC Diflorasone diacetate | 33564-31-7 | MFCD00079159 | 99.9% | 494.52 g/mol | C26H32F2O7 | 5 MG
SDP
Catalog No. 5000242448
Encompass_Preferred
Diflorasone diacetate is a synthetic topical corticosteroid used in research on corticosteroid-responsive dermatoses. Supplied as a white to off-white solid with high purity, it is intended for laboratory research use only and not for human or veterinary administration.
  • High purity suitable for analytical and pharmacological studies.
  • White to off-white solid form for easy handling and formulation.
  • Available in small milligram quantities for research applications.
  • Molecular weight 494.52 g/mol; formula C26H32F2O7.
  • Store sealed at 4°C; protect from moisture and light.
Catalog No. 50-002-42448 Supplier Medchemexpress LLC Supplier No. HY1079615MG
Diflorasone diacetate is a synthetic topical corticosteroid used in research on corticosteroid-responsive dermatoses. Supplied as a white to off-white solid with high purity, it is intended for laboratory research use only and not for human or veterinary administration.
  • High purity suitable for analytical and pharmacological studies.
  • White to off-white solid form for easy handling and formulation.
  • Available in small milligram quantities for research applications.
  • Molecular weight 494.52 g/mol; formula C26H32F2O7.
  • Store sealed at 4°C; protect from moisture and light.