Learn More
Medchemexpress LLC Difloxacin-d3 | 1173147-93-7 | 98.0% | 1 MG
SDP
Catalog No. 5000232848
Encompass_Preferred
Difloxacin-d3 is the deuterium-labeled form of the fluoroquinolone difloxacin supplied for research use as a stable-isotope tracer and internal standard for analytical quantification and metabolic tracing.
  • Deuterium-labeled (d3) tracer suitable for LC-MS and GC-MS analysis.
  • High chemical purity (~98.0%), appropriate for analytical standards.
  • Solid form (white to off-white) with defined handling and storage guidance.
  • Small pack sizes enable economical use for calibration and method development.
Catalog No. 50-002-32848 Supplier Medchemexpress LLC Supplier No. HY121272AS21MG
Difloxacin-d3 is the deuterium-labeled form of the fluoroquinolone difloxacin supplied for research use as a stable-isotope tracer and internal standard for analytical quantification and metabolic tracing.
  • Deuterium-labeled (d3) tracer suitable for LC-MS and GC-MS analysis.
  • High chemical purity (~98.0%), appropriate for analytical standards.
  • Solid form (white to off-white) with defined handling and storage guidance.
  • Small pack sizes enable economical use for calibration and method development.