Learn More
Cayman Chemical Difloxacin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1173147-93-7, 1 mg
SDP
Catalog No. 502938681
Encompass_Preferred
An internal standard for the quantification of difloxacin by GC- or LC-MS
Catalog No. 50-293-8681 Supplier Cayman Chemical Supplier No. 276171 MG
An internal standard for the quantification of difloxacin by GC- or LC-MS