Learn More
Medchemexpress LLC Difloxacin-d3 hydrochloride | 1173021-89-0 | 99.0% | 5 MG
SDP
Catalog No. 5000233737
Encompass_Preferred
Difloxacin-d3 hydrochloride is the deuterium-labeled hydrochloride salt of the fluoroquinolone antibiotic difloxacin. Supplied as a solid analytical standard for research use, it is used as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses; deuterium substitution may alter pharmacokinetic and metabolic behavior relative to the unlabeled compound.
  • Deuterium-labeled tracer for quantitative analysis
  • High purity (99.0%) suitable for analytical standards
  • Available in small mg quantities for method development
  • Solid form simplifies storage and handling
  • For research use only; not for human use
Catalog No. 50-002-33737 Supplier Medchemexpress LLC Supplier No. HY121272AS15MG
Difloxacin-d3 hydrochloride is the deuterium-labeled hydrochloride salt of the fluoroquinolone antibiotic difloxacin. Supplied as a solid analytical standard for research use, it is used as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS analyses; deuterium substitution may alter pharmacokinetic and metabolic behavior relative to the unlabeled compound.
  • Deuterium-labeled tracer for quantitative analysis
  • High purity (99.0%) suitable for analytical standards
  • Available in small mg quantities for method development
  • Solid form simplifies storage and handling
  • For research use only; not for human use