Learn More
Selleck Chemical LLC Difloxacin hydrochloride S5041-25mg
SDP
Catalog No. 503112400
Encompass_Preferred
Difloxacin is a second-generation synthetic fluoroquinolone antimicrobial antibiotic used in veterinary medicine
Catalog No. 50-311-2400 Supplier Selleck Chemical LLC Supplier No. S504125MG
Difloxacin is a second-generation synthetic fluoroquinolone antimicrobial antibiotic used in veterinary medicine