Learn More
Medchemexpress LLC Difloxacin hydrochloride | 91296-86-5 | 100 MG
SDP
Catalog No. 5000418399
Encompass_Preferred
Difloxacin hydrochloride is a broad-spectrum antibacterial agent. It inhibits bacterial DNA gyrase and exhibits a concentration-dependent bactericidal effect by interfering with the activity of DNA gyrase and topoisomerase IV.
  • Inhibits bacterial DNA gyrase
  • Exhibits a concentration-dependent bactericidal effect
  • Interferes with DNA gyrase and topoisomerase IV activity
Catalog No. 50-004-18399 Supplier Medchemexpress LLC Supplier No. HYN7066100MG
Difloxacin hydrochloride is a broad-spectrum antibacterial agent. It inhibits bacterial DNA gyrase and exhibits a concentration-dependent bactericidal effect by interfering with the activity of DNA gyrase and topoisomerase IV.
  • Inhibits bacterial DNA gyrase
  • Exhibits a concentration-dependent bactericidal effect
  • Interferes with DNA gyrase and topoisomerase IV activity