Learn More
Cayman Chemical Diflubenzuron, Purity: ≥98%, CAS: 35367-38-5, 100 mg
SDP
Catalog No. 502995542
Encompass_Preferred
An insecticide; inhibits chitin synthesis (IC50 = 0.611 nM); inhibits chitin synthetase at the egg and larval stages; genotoxic and mutagenic in mice at doses of 0.3, 1, and 3 mg/kg
Catalog No. 50-299-5542 Supplier Cayman Chemical Supplier No. 23095100 MG
An insecticide; inhibits chitin synthesis (IC50 = 0.611 nM); inhibits chitin synthetase at the egg and larval stages; genotoxic and mutagenic in mice at doses of 0.3, 1, and 3 mg/kg