Learn More
Medchemexpress LLC Diflubenzuron | 35367-38-5 | 98.7% | 50 MG
SDP
Catalog No. 5000410333
Encompass_Preferred
Diflubenzuron is an insecticide that primarily exhibits larvicidal and ovicidal activities. It functions by inhibiting the synthesis of chitin in insects, which is crucial for their exoskeleton development. This interference leads to defects in insect molting, ultimately causing their death. The product is intended for research use only and is not sold to patients.
  • Inhibits chitin synthesis in insects.
  • Affects insect molting.
  • Exhibits larvicidal activity.
  • Exhibits ovicidal activity.
Catalog No. 50-004-10333 Supplier Medchemexpress LLC Supplier No. HYB197350MG
Diflubenzuron is an insecticide that primarily exhibits larvicidal and ovicidal activities. It functions by inhibiting the synthesis of chitin in insects, which is crucial for their exoskeleton development. This interference leads to defects in insect molting, ultimately causing their death. The product is intended for research use only and is not sold to patients.
  • Inhibits chitin synthesis in insects.
  • Affects insect molting.
  • Exhibits larvicidal activity.
  • Exhibits ovicidal activity.