Learn More
Cayman Chemical DIflucortolon 21-valerate 10mg
SDP
Catalog No. 502948057
Encompass_Preferred
A glucocorticoid; has anti-inflammatory effects in a rat model of croton oil-induced EAr edema when applied topically (ED50 = 9.7 UG/ml); induces skin inflammation and thins the epidermis in hairless dogs following chronic topical administration at 0.1%; increases allergic- and non-allergic-mediated skin inflammation in mouse models of atopic dermatitis when applied topically prior to and following sensitization
Catalog No. 50-294-8057 Supplier Cayman Chemical Supplier No. 3320310 MG
A glucocorticoid; has anti-inflammatory effects in a rat model of croton oil-induced EAr edema when applied topically (ED50 = 9.7 UG/ml); induces skin inflammation and thins the epidermis in hairless dogs following chronic topical administration at 0.1%; increases allergic- and non-allergic-mediated skin inflammation in mouse models of atopic dermatitis when applied topically prior to and following sensitization