Learn More
Cayman Chemical Diflucortolone 21-valerate, Purity: ≥98%, CAS: 59198-70-8, 50 mg
SDP
Catalog No. 502948089
Encompass_Preferred
A glucocorticoid; has anti-inflammatory effects in a rat model of croton oil-induced ear edema when applied topically (ED50 = 9.7 µg/ml); induces skin inflammation and thins the epidermis in hairless dogs following chronic topical administration at 0.1%; increases allergic- and non-allergic-mediated skin inflammation in mouse models of atopic dermatitis when applied topically prior to and following sensitization
Catalog No. 50-294-8089 Supplier Cayman Chemical Supplier No. 3320350 MG
A glucocorticoid; has anti-inflammatory effects in a rat model of croton oil-induced ear edema when applied topically (ED50 = 9.7 µg/ml); induces skin inflammation and thins the epidermis in hairless dogs following chronic topical administration at 0.1%; increases allergic- and non-allergic-mediated skin inflammation in mouse models of atopic dermatitis when applied topically prior to and following sensitization