Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diflufenican | 83164-33-4 | MFCD01311804 | 99.8% | 394.29 g/mol | C19H11F5N2O2 | 1 ML
SDP

Catalog No. 5000208699
Encompass_Preferred

Diflufenican is a contact, selective herbicide used to control certain broadleaf weeds. Supplied for research use, it is commonly provided as a DMSO solution (example: 10 mM, 1 mL). Research applications include biological assays and analytical standards.

  • Solubility: DMSO: 66.67 mg/mL (169.09 mM); in vivo: ≥ 2.5 mg/mL in 10% DMSO/90% corn oil.
  • Purity: 99.77% (typical).
  • Formula and weight: C19H11F5N2O2; 394.29 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: research reagent; not for human or clinical use.

Catalog No. 50-002-08699 Supplier Medchemexpress LLC Supplier No. HYW04020610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diflufenican is a contact, selective herbicide used to control certain broadleaf weeds. Supplied for research use, it is commonly provided as a DMSO solution (example: 10 mM, 1 mL). Research applications include biological assays and analytical standards.

  • Solubility: DMSO: 66.67 mg/mL (169.09 mM); in vivo: ≥ 2.5 mg/mL in 10% DMSO/90% corn oil.
  • Purity: 99.77% (typical).
  • Formula and weight: C19H11F5N2O2; 394.29 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: research reagent; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.