Learn More
Medchemexpress LLC Diflunisal-d3 | 1286107-99-0 | 99.7% | 253.22 | 2.5 MG
SDP
Catalog No. 5000387580
Encompass_Preferred
Diflunisal-d3 is the deuterium labeled form of Diflunisal, a salicylate derivative with nonsteroidal anti-inflammatory and uricosuric properties. It acts as a Cyclooxygenase (COX) inhibitor and is used as an analgesic and in rheumatoid arthritis patients. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-87580 Supplier Medchemexpress LLC Supplier No. HY18342S2.5MG
Diflunisal-d3 is the deuterium labeled form of Diflunisal, a salicylate derivative with nonsteroidal anti-inflammatory and uricosuric properties. It acts as a Cyclooxygenase (COX) inhibitor and is used as an analgesic and in rheumatoid arthritis patients. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs.