Learn More
Cayman Chemical DIfluorInated H2S FluorScent
SDP
Supplier:  Cayman Chemical 29835500 UG
Encompass_Preferred
A fluorescent probe for H2S; ex/em = 365/450 nm, respectively; selectively fluoresces in the presence of H2S over Zn2+, Fe3+, S2O32-, ClO-, SO32-, H2O2, NO2-, Cys, Hcy, and GSH at 1 µM
Catalog No. 50-299-0539