Learn More
Cayman Chemical DIfluorInated H2S FluorScent
SDP
Catalog No. 502988247
Encompass_Preferred
A fluorescent probe for H2S; ex/em = 365/450 nm, respectively; selectively fluoresces in the presence of H2S over Zn2+, Fe3+, S2O32-, ClO-, SO32-, H2O2, NO2-, Cys, Hcy, and GSH at 1 µM
Catalog No. 50-298-8247 Supplier Cayman Chemical Supplier No. 29835250 UG
A fluorescent probe for H2S; ex/em = 365/450 nm, respectively; selectively fluoresces in the presence of H2S over Zn2+, Fe3+, S2O32-, ClO-, SO32-, H2O2, NO2-, Cys, Hcy, and GSH at 1 µM