Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Difluoroacetic Acid
Spécifications
Spécifications
| CAS | 381-73-7 |
| Melting Point | -1°C |
| Density | 1.526 g/mL at 25°C |
| Boiling Point | 132°C to 134°C |
| Molecular Formula | C2H2F2O2 |
| Refractive Index | n20/D 1.344 |
| Linear Formula | F2CHCO2H |
| MDL Number | MFCD00004220 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 96.03 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'